Tlačiť
DNase I Digest Kit, peqGOLD
PEQL12-1091-01EM
DNase I Digest Kit, peqGOLD
For quick and easy removal of unwanted DNA directly on the column during RNA preparation.
  • Efficient removal of contaminating DNA from RNA preparations
  • Free from RNases and inhibitors
Informace o dodávce: Kit contains RNase-free DNase I and DNase I digestion buffer.