Tlačiť
Gel Extraction Kit, peqGOLD
PEQLEMR501005EA 26.84 EUR
PEQL12-2501-02 PEQL12-2501-01 PEQL12-2501-00 PEQLEMR501200 PEQLEMR501005
Gel Extraction Kit, peqGOLD
Gel Extraction Kit for easy and time-saving isolation of DNA fragments from agarose gels, with high recovery rates up to 30 µg for fragments from 50 bp up to 40 kbp.
  • Simple protocol and short hands-on times of less than 15 minutes
  • Internal control of reaction conditions by pH sensitive indicator
  • No need for organic extractions or precipitations
  • Efficient removal of inhibitors and contaminants
  • No shearing of DNA or carry-over of silica material
  • Elution in TE buffer or water
Informace o dodávce: Kit contains PerfectBind DNA columns (Safety or Classic Line) and collection tubes, Binding Buffer with pH Indicator, CG Wash and Elution Buffer.