Tlačiť
dNTP set, long range, peqGOLD
732-3175EA 0 EUR
PEQL20-2110 PEQL20-2111
dNTP set, long range, peqGOLD
Designed for the successful PCR amplification of long DNA fragments, the peqGOLD dNTP set, long range contains one vial each of dATP, dCTP, dGTP and dTTP each at a concentration of 100 mM.