Tlačiť
dNTP mix, long range, peqGOLD
732-3182EA 0 EUR
PEQL20-3111
dNTP mix, long range, peqGOLD
Designed for the successful PCR amplification of long DNA fragments, the peqGOLD dNTP mix, long range contains premixed dATP, dCTP, dGTP and dTTP, ready-to-use, each at a concentration of 10 mM. Available in 3 quantities 2 µmol, 10 µmol and 50 µmol of each individual nucleotide, ie. 8 µmol, 40 µmol and 200 µmol mixed dNTPs.