Tlačiť
Agarose, peqGOLD MoSieve™ Agarose
Dodávateľ: Peqlab brand
732-3191EA 0 EUR
PEQL35-4030 PEQL35-3020 PEQL35-4010 PEQL35-3010
Agarose, peqGOLD MoSieve™ Agarose
peqGOLD MoSieve™ Agarose is a specialist agarose for separating small nucleic acid fragments.
  • High separation and band sharpness
  • Easily soluble, free from foaming
  • Time saving alternative to polyacrylamide gels