Tlačiť
Agarose, MegaBase™, peqGOLD
732-3193EA 0 EUR
PEQL35-5020
Agarose, MegaBase™, peqGOLD
peqGOLD MegaBase™ agarose is suitable for work with large DNA fragments, between 200 bp and 50 kbp in size. MegaBase™ possesses very high gel strength making it suitable for intense, short running times.
  • High separation and band sharpness
  • High optical transparency
  • Optimal solubility through autoclaving