Tlačiť
Automatic Kjeldahl Analyser, UDK 169
Dodávateľ: VELP Scientifica
709-0139EA 0 EUR
709-0139 709-0140 709-0141
Automatic Kjeldahl Analyser, UDK 169
Destilační systémy Kjeldahlův destilační systém
The UDK 169 distillation and titration system is designed to perform nitrogen and protein determination according to the Kjeldahl Method (TKN) in the food and feed industries and several other applications in environmental control (phenols, nitrogen in water, sludge, soil and lubricant) and also in the chemical and pharmaceutical industries after having digested the sample accurately.
  • Programmable boric acid, water, sodium hydroxide addition
  • Not necessary to set distillation time when titrating
  • Incorporates an AOAC recommended colorimetric titrator
  • Selectable steam generation output level 10 to 100%
  • Auto removal of residues from titrator and sample tube
  • Library of 55 programs (31 pre-defined, 24 customisable)
Designed with a highly resistant technopolymer housing for corrosion protection, the system is built around the tough, 'zero pressure' steam generator, titanium condenser and technopolymer splash head ensure the ultimate in nitrogen analysis for precise determination. High precision pumps and burette ensure constant accurate dosing of reagents.
The UDK distillation and titration system offers instantaneous reporting, powerful data handling with on-board archive for data storage, balance data input, output to PC, pen drive, printer in a common format, via Ethernet and USB.
UDK 169 with AutoKjel
Automated, safe operation with maximised productivity
Continuous process for 24×250 ml or 21×400 ml samples
Charge and discharge tubes with sensor
Unattended operation
Informace o dodávce: UDK 169</bold> is supplied with test tube Ø 42×300 mm; collecting flask 250 ml, pincer for test tubes, touch pen and protective film for touch screen.
<bold>AutoKjel autosampler</bold> comes complete with all the accessories required for the connection with UDK 169 and tanks for NaOH (10 litre), H₂O (20 litre), H₂BO₂ (5 litre) and waste (20 litre).