Tlačiť
Test sieves, 200×50 mm
Dodávateľ: Endecotts
ENDE200BAW31.5EA 271.98 EUR
ENDE200BAW31.5 ENDE200BAW.075 ENDE200BAW16.0 ENDE200BAW.032 ENDE200BAW.038 ENDE200BAW75.0 ENDE200BAW.355 ENDE200BAW.710 ENDE200BAW37.5 ENDE200BAW6.30 ENDE200BAW106. ENDE200BAW50.0 ENDE200BAW2.36 ENDE200BAW.150 ENDE200BAW90.0 ENDE200BAW25.0 ENDE200BAW.045 ENDE200BAW.125 ENDE200BAW.600 ENDE200BAW2.80 ENDE200BAW1.70 ENDE200BAW2.00 ENDE200BAW63.0 ENDE200BAW11.2 ENDE200BAW13.2 ENDE200BAW.053 ENDE200BAW53.0 ENDE200BAW.212 ENDE200BAW.850 ENDE200BAW19.0 ENDE200BAW1.40 ENDE200BAW.090 ENDE200BAW.250 ENDE200BAW4.75 ENDE200BAW1.00 ENDE200BAW22.4 ENDE200BAW.020 ENDE200BAW.500 ENDE200BAW45.0 ENDE200BAW1.18 ENDE200BAW.025 ENDE200BAW.300 ENDE200BAW.425 ENDE200BAW.106 ENDE200BAW26.5 ENDE200BAW9.50 ENDE200BAW.180 ENDE200BAW3.35 ENDE200BAW.063 ENDE200BAW125.
Test sieves, 200×50 mm
Síta Síta
These test sieves have a brass frame and stainless steel mesh.
Certifikácie: According to ASTM E 11-95
Informace o dodávce: Each sieve is delivered with an individual quality certificate.