Tlačiť
Rotational viscometers, DV1400
Dodávateľ: Industrial Physics
SIMEDV1401EA 3099.96 EUR
SIMEDV1401
Rotational viscometers, DV1400
These rotational viscometers are according to Brookfield method, for the quick determination of viscosity in the laboratory, research centres and production.
  • Two lines digital display with backlight
  • Compatible with standard rotational Brookfield viscometers
  • Accuracy ±2,0% of full scale range with repeatability of ±1,0%
The intuitive, easy functionality, light weight and battery operation provides a great versatility.
Informace o dodávce: Supplied with standard R2-R7 spindles, beakers, assembly tools, alkaline batteries and carrying case.