Tlačiť
Automated pipetting system, epMotion® M5073c
Dodávateľ: Eppendorf
EPPE5073000.400EA 0 EUR
EPPE5073000.400
Automated pipetting system, epMotion® M5073c
Automated Liquid Handling Systems
The M5073 automated pipetting systems is tailored for nucleic acid purification (NAP), but is flexible for other liquid handling tasks. The integrated magnetic finger and ThermoMixer module is ideal for our MagSep reagent kits, saving hands-on time and improving the reproducibility of results. The M5073 is controlled by EasyCon tablet and can be configured with CleanCap.
  • 6 ANSI/SLAS microplate positions
  • Compact control tablet with touchscreen or mouse operation, USB data transfer
  • Software assistants for simple, step-by-step application programming
  • Optical sensor for detecting liquids, labware, tips
  • Volume range of 1 to 1000 μl for maximum pipetting accuracy
  • 3D-MagSep technology, mixing, tempering, and magnetic separation all at one location
  • Calibrated 1-channel and 8-channel pipetting tools
  • UV lamp and HEPA filter system for decontamination and clean air conditions
  • Compatible with tubes (0,1 to 50 ml) and microplates with up to 384 wells
The epMotion M5073 features 2 systems in one. First, a customized system for automated nucleic acid purification of 1 to 24 samples with special MagSep reagents. The Eppendorf EasyCon tablet and Prep Assistant software provide simple operation with step-by-step guidance through the process. Secondly, a flexible system for automating time-consuming and complex pipetting procedures with the epBlue software, making routine Liquid Handling tasks easier than ever. The pipetting procedure is more precise, reproducible and fully standardized, making your workplace more ergonomic and safer. The epMotion M5073 is delivered with all accessories required for nucleic acid purification; only consumables need to be ordered separately.
Informace o dodávce: Supplied with software epBlue and Prep-Assistant; EasyCon tablet; CleanCap; dispensing tools TS 50, TS 1000, PrepRack; reagent rack; rack for 24 Safe-Lock tubes.