Tlačiť
Ashing furnaces with fold down or lift door, LV and LVT series
Dodávateľ: Nabertherm
455-1944EA 4350.76 EUR
455-1944 455-1950 455-1974 455-1986 455-1998 455-1938 455-2010 455-2016 455-1968 455-1962 455-1992 455-2008
Ashing furnaces with fold down or lift door, LV and LVT series
Sušárny Pece vysokoteplotní
These models are especially designed for ashing in the laboratory. Units feature rust-free, textured stainless steel exteriors. A special air intake and exhaust system allows air exchange of more than 6 times per minute. Incoming air is pre-heated to ensure a good temperature uniformity. Models have Tmax 1100 °C, and feature a silent solid state power control relay.
  • Heating on two sides by means of ceramic heating plates, module lining is highly durable, high performance cured vacuum fibre
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and is easy to replace
  • Double walled casing for low external temperatures and stability
  • Optional fold down door (LV models) which can be used as work platform or lift door (LVT models) with hot surface facing away from the operator
  • Air exchanged more than 6 times per minute
  • Non-classified insulation avoids the potential risk of cancer
Informace o dodávce: Supplied with Standard B 510 controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions or optional C 550 controller with LC display, 10 freely adjustable programs and 20 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. All units are supplied with plugs and these models require a 230 V single phase power supply.
Pozor: Maximum continuous operating temperature should be 50 °C below the max. temp. of the model shown in the table above.