Chcete sa opäť prihlásiť?
Gel documentation systems, VWR® Smart3 and Smart3 EZ
Dodávateľ: VWR Collection
VWR®
VWR® Smart3 and Smart3 EZ are complete stand-alone systems with integral processor, so no external PC is required.
  • Real time images from 12-/16-bit, 3 megapixel camera
  • Images can be saved as TIFF, BMP or JPEG files on a USB stick, hard drive or directly to a network via PC
  • Integrated touch screen with icon-driven menus to guide user through system functions
  • Sliding transilluminator tray offers easy access
  • Filter drawer with optional interchangeable filters enables viewing of a wide range of fluorescent stains
  • UV to white light converter screen (optional) simplifies imaging of protein gels, autoradiographs and colony plates
Using the VWR® Smart3 or Smart3 EZ system it is possible to produce images of electrophoresis gels stained with many many fluorescent and colorimetric dyes, including: Coomassie® blue, silver stain, ethidium bromide, SYBR® Gold, SYBR® Green, SYBR® Safe, GelStar®, SYPRO® Red, SYPRO® Ruby, SYPRO® Orange, Fluorescein, Pro-Q® Diamond, or Deep Purple™. As new dyes are released we work to optimise their use with the VWR® Smart systems, so please ask your local VWR contact for updates.
The VWR® Smart systems are also suitable for viewing and capturing images from agar plates of dark, light or two colour colonies, cells in flasks, autoradiographs, DNA, RNA or protein on membranes, spot and slot blots of DNA, RNA or protein, cells or solutions in microtitre plates and DNA or protein macroarrays.
Informace o dodávce: Complete system includes 3 megapixel 12-/16-bit camera, manual zoom lens (VWR® Smart3) or motor-driven zoom lens (VWR® Smart3 EZ), darkroom, network connection, built-in processor (no PC required), VWR® Image Capture Software and two copies VWR® Gel Documentation Software. Illumination options must be ordered separately.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
VWR® Smart3 1 730-1459
VWR® Smart3 EZ 1 730-1460
Transilluminators for use with VWR® Smart3
Opis Rozmery Pk Číslo v katalógu VWR Množstvo
UV to white light converter screen, 200×200 mm - 1 730-1395
UV to blue light conversion screen 210×260 mm - 1 730-1493
UV transilluminator (200×200 mm, 254 nm) 241×315×80 mm 1 730-1463
UV transilluminator (200×200 mm, 302 nm) 241×315×80 mm 1 730-1461
UV transilluminator (200×200 mm, 365 nm) 241×315×80 mm 1 730-1462
VWR® Blue slim transilluminator (100×120 mm, 470 nm) - 1 730-1468
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Model VWR® Smart3 EZ VWR® Smart3
Kamera 1/3" sensor
File bit depth (A/D) 12/16
Camera resolution (pixels) 3 megapixels
Dynamic range 3,6 - 4,8
Max. gel size, with hood (mm) 200×200
ŠxH×V (mm) 465×390×510
DOPLNKY
Rozšíriť všetko / Schovať všetko
Accessories
Opis Pk Číslo v katalógu VWR Množstvo
Thermal printer, digital 1 730-1260
Thermal paper, matt 1 Roll 730-2892
Thermal paper, glossy 1 Roll 733-2000
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo