Tlačiť

Anti-MS4A1 Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB05140-BEA 361.99 EUR
EVBIEB05140-B
Anti-MS4A1 Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 35kDa band observed in lysates of cell line Daudi (calculated MW of 33.1kDa according to NP_068769.2). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: MS4A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR