Tlačiť

Anti-GRB2 Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB05284-BEA 361.99 EUR
EVBIEB05284-B
Anti-GRB2 Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 26kDa band observed in lysates of Human Thymus and of cell line MOLT4 (calculated MW of 25.2kDa according to NP_002077.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: GRB2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR