Tlačiť

Anti-ANXA11 Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB05307-BEA 362.77 EUR
EVBIEB05307-B
Anti-ANXA11 Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 55kDa band observed in Jurkat lysates (calculated MW of 54.4kDa according to NP_001148.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: ANXA11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR