Tlačiť

Anti-GAPDH Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB06377-BEA 361.99 EUR
EVBIEB06377-B
Anti-GAPDH Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 35kDa band observed in lysates of cel line HEK293 (calculated MW of 36.1kDa according to NP_002037.2). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Dog, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR