Tlačiť

Anti-APOBEC3G Goat Polyclonal Antibody

Dodávateľ: Everest Biotechnology
EVBIEB06714EA 362.77 EUR
EVBIEB06714
Anti-APOBEC3G Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: APOBEC3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR