Tlačiť

Anti-SERPINE1 Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB06720-BEA 361.99 EUR
EVBIEB06720-B
Anti-SERPINE1 Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 47kDa band observed in lysates of cell line HepG2 (calculated MW of 45.1kDa according to NP_000593.1). The observed molecular weights correspond to earlier findings with different antibodies from other commercial sources. See non-bio

Type: Primary
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR