Tlačiť
Anti-PVALB Goat Polyclonal Antibody (Biotin)
Dodávateľ: Everest Biotechnology
EVBIEB06776-BEA 361.99 EUR
EVBIEB06776-B
Anti-PVALB Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 12kDa band observed in Human Brain (Cerebellum) lysates (calculated MW of 12.1kDa according to NP_002845.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: PVALB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat