Tlačiť

Anti-SDHB Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB06868-BEA 361.99 EUR
EVBIEB06868-B
Anti-SDHB Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 28kDa band observed in Human Liver lysates (calculated MW of 31.6kDa according to NP_002991.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR