Tlačiť

Anti-CBX5 Goat Polyclonal Antibody (Biotin)

Dodávateľ: Everest Biotechnology
EVBIEB06957-BEA 361.99 EUR
EVBIEB06957-B
Anti-CBX5 Goat Polyclonal Antibody (Biotin)
Protilátky
Western Blot: Approx 25kDa band observed in HeLa lysates (calculated MW of 22.2kDa according to NP_036249.1). See non-biotinylated parental product's datasheet for further QC data. Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: CBX5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR