Tlačiť

Anti-AURKB Goat Polyclonal Antibody

Dodávateľ: Everest Biotechnology
EVBIEB07260EA 361.99 EUR
EVBIEB07260
Anti-AURKB Goat Polyclonal Antibody
Protilátky
Western Blot: Approx 37kDa band observed in Human Thymus lysates and approx. 35kDa in Human Lymph Node, Spleen, Tonsil and Testis lysates (calculated MW of 39.3kDa according to NP_004208.2). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR