Tlačiť

Anti-LGALS3 Goat Polyclonal Antibody

Dodávateľ: Everest Biotechnology
EVBIEB07710EA 362.77 EUR
EVBIEB07710
Anti-LGALS3 Goat Polyclonal Antibody
Protilátky
Western Blot: Approx 30kDa band observed in Human and Mouse Colon lysates (calculated MW of 26.2kDa according to NP_002297.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Lotz et al, Proc Natl Ac

Type: Primary
Antigen: LGALS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR