Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-BMPR2 Goat Polyclonal Antibody

Dodávateľ: Everest Biotech
Western Blot: Approx 110kDa band observed in Human Liver lysates (calculated MW of 115kDa according to NP_001195.2). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Bovine




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Pack type Číslo v katalógu VWR Množstvo
100µg 0,1 mg plastic vial EVBIEB08820
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol BMPR2
Antigen name bone morphogenetic protein receptor, type II (serine/threonine kinase)
Reactivity Dog, Mouse, Human, Bovine, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Antigen synonyms BRK-3, POVD1, T-ALK, BMPR3, PPH1, BMPR-II, BMR2
Gene ID 659
Concentration 100µg/200µl
Format affinity purified
Purification Affinity purified
Sequence C-DLETNKLDPKEVDK
Form liquid
Storage buffer Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
Storage temperature Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
Shipping temperature 8 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo