Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-MYH9 Goat Polyclonal Antibody

Dodávateľ: Everest Biotech
Western Blot: Approx 250kDa band observed in lysates of Human Peripheral Blood Mononucleocytes and of NIH3T3 (calculated MW of 227kDa according to Human NP_002464.1 and to Mouse NP_071855.2). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: MYH9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Pack type Číslo v katalógu VWR Množstvo
100µg 0,1 mg plastic vial EVBIEB09020
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol MYH9
Antigen name myosin, heavy chain 9, non-muscle
Reactivity Human, Rat, Dog, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Antigen synonyms NMMHCA, NMHC-II-A, BDPLT6, FTNS, DFNA17, NMMHC-IIA, EPSTS, MHA
Gene ID 4627
Concentration 100µg/200µl
Format affinity purified
Purification Affinity purified
Sequence C-DQINTDLNLERSH
Form liquid
Storage buffer Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
Storage temperature Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
Shipping temperature 8 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo