Tlačiť
Multi-parameter meter, bench, ProLab 2500
Dodávateľ: SI Analytics
663-0316EA 3225.01 EUR
663-0316 663-0332 663-0331 663-0333
Multi-parameter meter, bench, ProLab 2500
Meřiče elektrochemické Meřiče pH a multimetry
The ProLab 2500 is a digital high-performance meter with three channels for connecting pH, ORP, ISE, dissolved oxygen, or conductivity IDS® sensors. With its colour graphic display protected by glass and the high-quality zinc die-casting base it meets even the highest demands.
  • Universal channels allow any combination of the same or different parameters
  • Digital sensor recognition: Digital signal transfer eliminates interference, safely allocates calibration data, and automatically transmits sensor data
  • Quality Sensor Control (QSC) icon provides precise information about the actual state of the electrode and therefore increases the operational safety
  • CMC function for monitoring the congruency of measuring and calibration range for pH
  • Data output to PC, USB memory stick or selected printers
  • Antibacterial keyboard
Informace o dodávce: Meter only is supplied with Z600 IDS® adapter for conventional pH electrodes (DIN plug), universal power supply and stand.
ProLab 2500 pH Set additionally includes A 162 IDS® pH sensor and buffer solutions.
ProLab 2500 pH/Cond Set additionally includes A 162 IDS® pH sensor, LF 413T IDS® conductivity cell, and testing solutions.
ProLab 2500 pH/Cond/Ox Set additionally includes A 162 IDS® pH sensor, LF 413T IDS® conductivity cell, FDO 1100 IDS® optical DO sensor, and testing solutions.