Tlačiť

Sucrose, 80%, in MNE buffer, MES + NaCl + EDTA

Dodávateľ: Thermo Fisher Scientific
J60720.APEA 93.69 EUR
J60720.AP
Sucrose, 80%, in MNE buffer, MES + NaCl + EDTA
Pufry Pufry extrakční
Physical Form: Viscous liquid
MDL No.: MFCD00006626
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR