Tlačiť

TRIS-CAPS buffer (10X)

Dodávateľ: Thermo Fisher Scientific
J61339.K2EA 138.83 EUR
ALFAJ61339.K2
TRIS-CAPS buffer (10X)
Physical Form: Liquid
Applications: For blot transfer
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR