Tlačiť

Succinate, 0,2 M buffer solution, pH 6,0

Dodávateľ: Thermo Fisher Scientific
J61853.APEA 94.5 EUR
J61853.AP J61853.AK
Succinate, 0,2 M buffer solution, pH 6,0
Pufry Pufry biologické
CAS No.: 150-90-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002789
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR