Tlačiť
Orange G/Blue dyes (6X)
Dodávateľ: Thermo Fisher Scientific
J62098.ACEA 69.19 EUR
ALFAJ62098.AC ALFAJ62098.AB
Orange G/Blue dyes (6X)
Orange G/Blue dye (6X) containing 0,25% orange-G, 0,25% bromophenol blue, 0,25% xylene cyanol FF, 30 mM EDTA in autoclaved DI water.