Tlačiť

MOBS, 0,2M buffer soln., pH 8,5

Dodávateľ: Thermo Fisher Scientific
J62203.AKEA 81.73 EUR
ALFAJ62203.AK
MOBS, 0,2M buffer soln., pH 8,5
CAS No.: 115724-21-5
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00674916
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR