Tlačiť
Electrode stirrer stand
Dodávateľ: Hach Lange
HACH4530002
Electrode stirrer stand
This magnetic stirrer and electrode stand holds up to three electrodes with adjustable stop. Will not heat sample.