Tlačiť
Human somatic stem cell media, HyClone AdvanceSTEM™
Dodávateľ: HyClone products (Cytiva)
HYCLSH30886.02EA 285.14 EUR
HYCLSH30886.02
Human somatic stem cell media, HyClone AdvanceSTEM™
Média kultivační
HyClone AdvanceSTEM™ Human Somatic Stem Cell Media has been developed for mesenchymal stem cell research to support applications ranging from the expansion and maintenance of human adult mesenchymal stem cells, to the directed differentiation of these cells into such cell types as neurons, adipocytes, chondrocytes and osteocytes.
HyClone AdvanceSTEM™ Adipogenic differentiation medium supports adipogenic differentiation from human adipose-derived and bone marrow-derived mesenchymal stem cells.
HyClone AdvanceSTEM™ Chondrogenic differentiation medium supports differentiation of a variety of hMSCs into chondrocytes.
HyClone AdvanceSTEM™ Osteogenic differentiation medium supports the differentiation of a variety of human mesenchymal stem cells including human adipose-derived, multipotent cord.