Tlačiť
Streptavidin coated plate IP kit, Pierce™
Dodávateľ: Thermo Fisher Scientific
Pierce
PIER45360EA 621.34 EUR
PIER45360
Streptavidin coated plate IP kit, Pierce™
Systémy pro purifikaci proteinů Protein Purification Kits
Pierce™ Streptavidin Coated Plate IP Kit uses coated streptavidin microplates to perform immunoprecipitation assays in 96-well plates without resins, beads, centrifugation or magnets.
  • Ready to use, high quality coated plates provide high capacity and consistency
  • Plate format best suited for simultaneously processing multiple samples and their control conditions
  • Faster, easier and more thorough washing than with traditional tube/resin IP methods
Informace o dodávce: Kit includes streptavidin coated 96-well strip plates (2 plates), uncoated 96-well strip plate, white (2 plates), PBS pack (makes 500 ml), Biotin Blocking Buffer (2×15 ml), Surfact-Amps™ X-100 detergent solution (6×10 ml), elution buffer pH 2,8 (50 ml), neutralisation buffer (12 ml), and sealing tape for 96-well plates (18 sheets).