Tlačiť
Flip-top vials for quality testing of milk, 60 ml
Dodávateľ: Capitol Europe
CAPI03036079WRWLCNEA 0 EUR
CAPI03036079WRWLCN
Flip-top vials for quality testing of milk, 60 ml
Vialky
Single use vials made of pure PP with rubber top suitable for food contact and used for quality testing of millk on automatic samplers.
  • Manufactured in a cleanroom and proprietary 'in mould closing' process
  • Perfectly tight and clean
  • Aseptic and 100% recyclable after use
  • Translucent in colour for visual inspection
  • One hand opening and closing for perfect cleanliness
Certifikácie: Complies with CE norm No. 1935/2004
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904