Tlačiť

Anti-V5 Tag Rabbit Polyclonal Antibody (CF543)

Dodávateľ: Biotium
20441.EA 472.51 EUR
20441.
Anti-V5 Tag Rabbit Polyclonal Antibody (CF543)
Protilátky
Polyclonal anti-V5 (GKPIPNPLLGLDST) reacts with V5 tagged fusion proteins. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™543 dye. CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Tag
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Rabbit
Isotype:
Reactivity: V5 tag
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR