Tlačiť
Fluor-free acids, DyLight™
Dodávateľ: Thermo Fisher Scientific
PIER62280EA 349.37 EUR
PIER62284 PIER62280 PIER62283
Fluor-free acids, DyLight™
Systémy pro spojování a značení proteinů
DyLight™ fluor-free acids are non-activated forms of selected DyLight™ fluorescent dyes for use as controls and calibration factors in fluorescence imaging applications. These non-activated fluorescent dyes contain the native carboxyl group of the standard dye molecule.