Thermo Scientific™ Versette™ is a user-friendly liquid handler in a compact format. It is compatible with 96- and 384-channel pipetting heads with a total volume range of 0,5 to 300 µl. Combined with DARTs tips, Versette liquid handler guarantees accurate and precise pipetting results in all a pplications across the entire pipetting range.
- Pipetting heads can be easily changed by the user without using any tools, while the process is safely guided step-by-step by Thermo Scientific™ ControlMate™ Software
- All pipetting heads utilise RFID tags for self-identity
- Compatible with D.A.R.T.s™ tips with unique sealing properties
- Six-position stage uses an innovative dual-level structure to minimise space requirements in the laboratory
- User-friendly programming options and visual icon-based on-board touchscreen
- Software provides a straightforward approach to creating and running pipetting programs, it includes a wide labware library and precise controlling options for various liquid handling applications
- Operating system: Windows XP SP3, Windows 7 (32- and 64-bit)
The Versette is designed to perform a variety of liquid handling tasks for a wide range of applications including plate stamping, plate reformatting, serial dilution and many other applications in low- to high-throughput laboratories. The instrument is also optimised for high-throughput immunoaffinity sample preparation with Mass Spectrometric Immunoassay (MSIA) pipette tips as part of the integrated MSIA workflow. The MSIA tips are compatible with the Versette where sample binding, wash and elution steps are automated to facilitate the cycling of samples and the appropriate buffers through the microcolumn embedded in the pipette tip. The complete walk-away immunoaffinity system provides increased sample throughput while providing high assay reproducibility.