Tlačiť
HyClone™ LS250 Lipid supplement
Dodávateľ: HyClone products (Cytiva)
HYCLSH30555.01EA 275.35 EUR
HYCLSH30555.01
HyClone™ LS250 Lipid supplement
HyClone™ LS250 Supplement is designed to stimulate cell growth and MAb production in NS0 cell cultures using traditional hybridoma serum-free media. LS250 Supplement has been successfully tested in a variety of media, including Cytiva HyClone™ CDM4MAb and SFM4Mab media.
  • Stimulates cell growth and monoclonal antibody production in cholesterol auxotrophic NS0 cell lines
  • Developed through Metabolic Pathway Design approach
  • Contains no animal-derived components (ADCF)
  • 250X concentrate formulated for enhanced stability and filterability
  • May be used to supplement serum-free media and in the preparation of liquid media media from dry powdered media