Tlačiť
Cryogenic Storage Systems, CryoExtra™
Dodávateľ: Thermo Scientific
THBRCE8120MEA 29057.79 EUR
THBRCE8120M THBRCE8180M THBRCE8140M THBRCE8194M
Cryogenic Storage Systems, CryoExtra™
Systémy pro kryoprezervaci
CryoExtra™ series high-efficiency storage vessels are designed to accommodate both vapor and liquid phase storage.
  • Sample capacity from 19500 up to 93000, 1,2 to 2,0 ml vials
  • Minimal footprint and real-time level monitor
  • Outstanding temperature uniformity and stable lid opening temperature
  • Advanced temperature monitor and alarms
  • Automated fill and level monitoring
  • De-fog switch for easy sample location
  • Hot gas bypass keeps warm nitrogen gas from impacting samples during a fill cycle
  • User comfort features and convenient, built-in workspace
  • Lockable lid for sample protection
  • Compatible with Thermo Scientific™ Smart-Vue™ wireless monitoring
Local power cords and plug sets are available.