Tlačiť
Chemical resistant ventilated suit, Hapichem™
Dodávateľ: Honeywell Safety
Hapichem™
113-8695EA 0 EUR
113-8695 113-8696 113-8693 113-8694 113-8692
Chemical resistant ventilated suit, Hapichem™
Jednorázové oděvy
This chemical resistant and anti-static suit is designed for short-duration operations, in areas where the air is polluted with solid particles and whole body protection is required. The suit is made of non woven laminated fabric with a barrier film. It is specially designed for breathable air network with low pressure. Compatible with decontamination showers.
  • High optical quality visor for excellent visibility and high chemical protection
  • Front zip for easy and quick donning and doffing (even when alone)
  • With valve: Airflow is adjustable to improve comfort
  • Integral back air ventilation improves comfort and enables users to sit down or to work on their knees
  • O-ring: Possibility to connect own gloves according to risks and workers sizes
  • Safety strip maximises safety for a quick emergency procedure
  • Halogen-free material is environmentally friendly - possibility to incinerate the suit after use
Double tight zip and sealed seams ensure the highest protection (Protection Factor ≥50000). Magnetic exhaust valve permits use in low pressure, to have more operators connected at the same time (low air consumption), and to ensure the highest protection.
Fabric thickness: 0,38 mm
Weight: <2 kg
Noise level: <75 dB at maximum flow rate
Minimum flow rate: 300 L/min in 2,8 bars of pressure network
Maximum flow rate: 730 L/min in 4,6 bars of pressure network.
Certifikácie: Protection factor Class 5 (>50000) according to EN 1073-1. Type 3 according to EN 14605. Surface resistivity <2,5×10e9 Ohm according to EN 1149-1. Cat. III