Tlačiť
Blotting membranes, nylon, TotalBLOT+™
Dodávateľ: VWR Life Science
TotalBLOT+™
E576-1ROLLEA 790.82 EUR
E576-1ROLL
Blotting membranes, nylon, TotalBLOT+™
Membrány transferové
TotalBLOT+™ positively charged nylon membranes are ideal for superior binding of negatively charged biomolecules in a wide variety of nucleic acid and protein applications. The surface chemistry is characterised by a high density of strongly cationic quaternary ammonium groups which are an integral part of the membrane.
  • Higher sensitivities and rapid nucleic acid binding without the need for UV crosslinking or baking