Tlačiť
PCR DNA Marker™ with loading dye
VWRCK811-50RXN
PCR DNA Marker™ with loading dye
PCR DNA Marker™ with loading dye offers 8 fragments ranging from 50 to 2000 bp. Marker is premixed with loading dye for added convenience. The marker contains high intensity reference bands that are ideal for quick sizing of PCR products and restriction digests. PCR DNA Marker™ with loading dye is ready to use, simply by loading 10 µl per lane. There is sufficient material for 50 assays.