Tlačiť
Crucibles
Dodávateľ: Thermo Fisher Scientific
35975.KTEA 530.17 EUR
35975.KT
Crucibles
Kelímky
These crucibles are manufactured from molybdenum. Available in straight wall type.
  • Excellent strength at high temperatures
  • High melting point of 2610 °C
  • High resistance to corrosion
  • Withstand repeated rough handling
Vessels of molybdenum can be used for such applications as processing nuclear fuel pellets at temperatures up to 1650 °C.