Tlačiť
Gas analysis crucibles
Dodávateľ: Thermo Fisher Scientific
39811.KNEA 167.34 EUR
39811.KN
Gas analysis crucibles
Kelímky
These crucibles are manufactured from graphite.
The crucibles for the analysis of gas content in metals are made from materials exclusively designed for this application. This material, along with a special post treatment of the machined crucible results in a vessel with exceptionally sensitive characteristics for analysing gases in metals.