Tlačiť
Lithium standard solution, 1000 µg/ml, BAKER INSTRA-ANALYZED®, J.T.Baker®
6456-04EA 0 EUR
BAKR6456-04
Lithium standard solution, 1000 µg/ml, BAKER INSTRA-ANALYZED®, J.T.Baker®
Lithium standard solution for AAS.
  • Packaged at a concentration of 1000 µg/ml in 5% nitric acid
  • Standardised with NIST standard chemicals
Prepared by dissolution of lithium carbonate of 99999% spectral purity. The metal content of the solution is confirmed by analysis, traceable to NIST SRM 3129a.
Balenie: Standard is packaged in a polyethylene bottle.