Tlačiť
Wash bottles, wide neck, non vented, LDPE, bleach, Azlon®
Dodávateľ: SciLabware
Azlon®
215-3858EA 48.16 EUR
215-3858
Wash bottles, wide neck, non vented, LDPE, bleach, Azlon®
Láhve Střičky
Multi-lingual wide neck wash bottle, made of opaque white LDPE for light sensitive sodium hypochlorite / bleach. Fitted with colour coded one-piece yellow PP closure.
  • Indelibly printed in three languages - English, French and Spanish
  • Features the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
  • Closure features precision jet-tip aperture for fine control