Tlačiť
Labels for BMP™ 71 printer, type B-342, PermaSleeve
Dodávateľ: Brady
PermaSleeve
BRDY115087EA 106.86 EUR
BRDY115087 BRDY115085 BRDY114958 BRDY114957 BRDY114979 BRDY114998 BRDY114942 BRDY115019 BRDY114978 BRDY115008 BRDY115009 BRDY115015 BRDY114985 BRDY115005 BRDY114984 BRDY115016 BRDY115003 BRDY115122 BRDY114980
Labels for BMP™ 71 printer, type B-342, PermaSleeve
Štítky
PermaSleeve® Heat-shrink polyolefin label provides a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Die-cut sleeves are cut to length
  • Contains the heat shrinkable wire marking sleeves
  • Supplies sold with the ribbon seperate, not in the same enclosure as the tape/sleeves
Certifikácie: Sleeves are self-extinguishing and meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, MIL-STD-202 method 215K and is UL 224 recognised. CSA acceptance pending.