Tlačiť
Ink-based printers, Orion™ PRT300, for Orion meters
Dodávateľ: Thermo Scientific
662-0893EA 1185.22 EUR
ORIOPRT301 ORIOPRT300
Ink-based printers, Orion™ PRT300, for Orion meters
This printer can be used with Thermo Scientific™ Orion™ Aplus and PerpHecT meters including 115A+, 125A+, 150A+, 145A+, 162A, 250A+, 290A+, 410A+, 420A+, 520A+, 525A+, 710A+, 720A+, 805A+, 810A+, 850A+, 920A+, 330, 350, 370, 550A, 535A, 555A and 862A.
  • Create printouts to meet GLP requirements
  • Ink-based
Objednávacie informácie: Supplied with ribbon, power cord, and Orion A+ and PerpHecT meter interface cable. Paper and ink cartridges need to be ordered separately.