Tlačiť
Wireless monitoring solution, RmoniWeb
Dodávateľ: Rmoni
Sensor2web™
620-2848EA 0 EUR
620-2848
Wireless monitoring solution, RmoniWeb
Systémy monitorování Systémy monitorování CO2 a teploty
A wireless monitoring system for permanent registration and surveillance of critical parameters. Typical applications include the temperature of food products inside cooling units and transport vehicles, environmental conditions in pharmaceutical cleanrooms, and the air quality in public buildings.
  • Wireless sensors carry out permanent and accurate measurements of critical parameters
  • All data are carefully and securely registered in a centralized datacenter
  • Data can be accessed via the internet from any PC
  • Ready-to-use and configurable reports provide regulatory authorities, customers, or internal auditors with all of the information they may require
  • Alerts via SMS and/or email enable users to act immediately when predefined threshold values have been exceeded
  • Independent of any brand or type of equipment: Ensures that investment can be re-used when replacing or repairing a refrigerating device or installation
The system enables close and comprehensive monitoring of different parameters and a rapid response in the event of deviations. With its unique combination of wireless technology and web-based software, it provides a cost-efficient and time-saving alternative to manual measurements.
A truly cloud-based software offering high availability for users. The platform is in constant evolution, adding features and capacity on a daily basis. The ideal choice for users who do not want to bother about servers, software updates, backups, UPS etc.
Certifikácie: Compliant to the standards EN 12830 and EN 13485.
Objednávacie informácie: All components of the system need to be ordered separately. Special kits including all required components are available separately.
For information regarding installation, calibration and other services, please contact your local Avantor customer service centre.
Please note: The monthly subscription is pre-invoiced, and cancellable via simple notice per quarter-end. Alarm SMS are post-invoiced or included in the yearly fee.